Gene Transcripts

Gene Symbol
Zfat
Entrez Gene ID
380993
Gene Name
zinc finger and AT hook domain containing
Gene Aliases
Gm922, Zfat1, Zfp406
Chromosome Location
Chr. 15: 68083714 - 68259265 on Build GRCm38
UniGene ID
Mm.309526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145888.1 NP_001139360.1 6 1050
NM_198644.2 NP_941046.1 6 1029
GenBank mRNA BC053930.1 - 6 1020
EU221277.1 - 6 880

View Full Product Details