Gene Transcripts

Gene Symbol
Fbxw24
Entrez Gene ID
382106
Gene Name
F-box and WD-40 domain protein 24
Gene Aliases
EG382106, Gm5162
Chromosome Location
Chr. 9: 109601116 - 109626059 on Build GRCm38
UniGene ID
Mm.441401
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013776.4 NP_001013798.2 4 361
GenBank mRNA BC080701.1 - 4 362
CF913977.1 - 4 346
CJ075113.1 - 4 361
GQ418172.1 - 4 362

View Full Product Details