Gene Transcripts

Gene Symbol
Enthd1
Entrez Gene ID
383075
Gene Name
ENTH domain containing 1
Gene Aliases
Gm1242, Gm86
Chromosome Location
Chr. 15: 80451721 - 80565157 on Build GRCm38
UniGene ID
Mm.297626
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163189.1 NP_001156661.1 1 383
GenBank mRNA BC158112.1 - 1 383

View Full Product Details