Gene Transcripts

Gene Symbol
Slc25a34
Entrez Gene ID
384071
Gene Name
solute carrier family 25, member 34
Gene Aliases
Gm1369
Chromosome Location
Chr. 4: 141618824 - 141623993 on Build GRCm38
UniGene ID
Mm.295682
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013780.1 NP_001013802.1 4 880
GenBank mRNA AK142924.1 - 4 882

View Full Product Details