Gene Transcripts

Gene Symbol
Fam47e
Entrez Gene ID
384198
Gene Name
family with sequence similarity 47, member E
Gene Aliases
C730043O17, Gm1381
Chromosome Location
Chr. 5: 92554951 - 92591285 on Build GRCm38
UniGene ID
Mm.76757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033478.2 NP_001028650.2 7 786
NM_001170571.1 NP_001164042.1 5 603
GenBank mRNA AK050399.1 - 3 304
AK144479.1 - 7 786
BC147167.1 - 3 284 9056206
BC147168.1 - 3 284 9056207
BE650192.1 - 6 611
DV648863.1 - 3 429

View Full Product Details