Gene Transcripts

Gene Symbol
Ano7
Entrez Gene ID
404545
Gene Name
anoctamin 7
Gene Aliases
Ngep, Ngep-L, Pcanap5, Tmem16g
Chromosome Location
Chr. 1: 93373796 - 93404304 on Build GRCm38
UniGene ID
Mm.297411
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271884.1 NP_001258813.1 23 2623
NM_207031.2 NP_996914.1 23 2623
GenBank mRNA BC116706.1 - 23 2589

View Full Product Details