Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012324.2 | NP_001012324.1 | 5 | 1207 | |
GenBank mRNA | AK038812.1 | - | 5 | 1069 | |
AK040995.1 | - | 5 | 1231 | ||
AK134746.1 | - | 5 | 1161 | ||
AK163501.1 | - | 4 | 3229 | ||
BC065151.1 | - | 5 | 1077 | 5708030 | |
BC089559.1 | - | 5 | 1207 | 30295705 |