Gene Transcripts
- Gene Symbol
- Zyg11b
- Entrez Gene ID
- 414872
- Gene Name
- zyg-ll family member B, cell cycle regulator
- Gene Aliases
- 1110046I03Rik, 2810482G21Rik, D4Mgi23, Gm431, mKIAA1730
- Chromosome Location
- Chr. 4: 108227755 - 108301141 on Build GRCm38
- UniGene ID
- Mm.426241
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033634.3 |
NP_001028806.2
|
10, 11 |
1862 |
|
GenBank mRNA |
AK122545.1 |
- |
3, 4 |
251 |
|
AK145130.1 |
- |
10, 11 |
1836 |
|
AK148335.1 |
- |
10, 11 |
1861 |
|
AK148760.1 |
- |
4, 5 |
332 |
|
AK166031.1 |
- |
10, 11 |
1842 |
|
AK166174.1 |
- |
10, 11 |
1842 |
|
AK166278.1 |
- |
10, 11 |
1853 |
|
BC139383.1 |
- |
10, 11 |
1829 |
8862405 |
BC139384.1 |
- |
10, 11 |
1829 |
8862406 |
View Full Product Details