Gene Transcripts

Gene Symbol
Nom1
Entrez Gene ID
433864
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
D5Kng1, Gm1040
Chromosome Location
Chr. 5: 29434659 - 29452169 on Build GRCm38
UniGene ID
Mm.262815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033457.2 NP_001028629.2 2 1035
GenBank mRNA AK148406.1 - 2 1039

View Full Product Details