Gene Transcripts

Gene Symbol
Vmn1r27
Entrez Gene ID
171206
Gene Name
vomeronasal 1 receptor 27
Gene Aliases
V1rc33
Chromosome Location
Chr. 6: 58215106 - 58216017 on Build GRCm38
UniGene ID
Mm.389781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134436.2 NP_602318.2 1 827

Gene Symbol
Vmn1r20
Entrez Gene ID
434017
Gene Name
vomeronasal 1 receptor 20
Gene Aliases
EG434017, Gm5569
Chromosome Location
Chr. 6: 57431691 - 57432602 on Build GRCm38
UniGene ID
Mm.473685
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101533.1 NP_001095003.1 1 827

View Full Product Details