Gene Transcripts
- Gene Symbol
- Zfp97
- Entrez Gene ID
- 22759
- Gene Name
- zinc finger protein 97
- Gene Aliases
- NGD16-4
- Chromosome Location
- Chr. 17: 17121383 - 17146878 on Build GRCm38
- UniGene ID
- Mm.436633
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011765.5 |
NP_035895.3
|
4 |
1472 |
|
GenBank mRNA |
L20450.1 |
- |
|
1440 |
|
- Gene Symbol
- Zfp960
- Entrez Gene ID
- 449000
- Gene Name
- zinc finger protein 960
- Gene Aliases
- Chromosome Location
- Chr. 17: 17064103 - 17089631 on Build GRCm38
- UniGene ID
- Mm.483718
- Species
- Mus musculus
View Full Product Details