Gene Transcripts

Gene Symbol
Ms4a15
Entrez Gene ID
545279
Gene Name
membrane-spanning 4-domains, subfamily A, member 15
Gene Aliases
E530011F12Rik, Gm1286
Chromosome Location
Chr. 19: 10978303 - 10993306 on Build GRCm38
UniGene ID
Mm.329729
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034898.2 NP_001030070.1 6, 7 691
GenBank mRNA AK143446.1 - 6, 7 692
BC147202.1 - 6, 7 688 9056250
BC172006.1 - 6, 7 752
BC172010.1 - 6, 7 766 9054296

View Full Product Details