Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145985.4 | NP_666097.3 | 2 | 185 | |
GenBank mRNA | AK028556.1 | - | 2 | 190 | |
AK031385.1 | - | 2 | 176 | ||
AK047995.1 | - | 2 | 223 | ||
AK128973.1 | - | 2 | 190 | ||
AK147747.1 | - | 2 | 220 | ||
AK154066.1 | - | 189 | |||
AK219764.1 | - | 1 | 15 | ||
BC017124.1 | - | 2 | 180 | 4480874 | |
BC023728.1 | - | 2 | 204 | 5351777 | |
BC033387.1 | - | 2 | 185 | 5368060 | |
BC034754.1 | - | 2 | 162 | 4954556 | |
BC083152.1 | - | 178 | 30461105 |