Gene Transcripts

Gene Symbol
Sult2a5
Entrez Gene ID
434264
Gene Name
sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 5
Gene Aliases
EG434264, Gm15438, OTTMUSG00000021844
Chromosome Location
Chr. 7: 13623967 - 13670807 on Build GRCm38
UniGene ID
Mm.359414
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184980.1 NP_001171909.1 1 158
GenBank mRNA AI315523.1 - 1 158

Gene Symbol
Sult2a4
Entrez Gene ID
434121
Gene Name
sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 4
Gene Aliases
EG434121, Gm5584
Chromosome Location
Chr. 7: 13909677 - 13989591 on Build GRCm38
UniGene ID
Mm.482762
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101534.1 NP_001095004.1 1 77

View Full Product Details