Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030694.1 | NP_109619.1 | 2 | 575 | |
GenBank mRNA | AK141392.1 | - | 2 | 602 | |
AK167865.1 | - | 2 | 313 | ||
AY082486.1 | - | 2 | 597 | ||
BC002102.1 | - | 2 | 575 | 3485116 | |
BC002160.1 | - | 2 | 580 | 3485614 | |
BC084679.1 | - | 2 | 575 | 5374182 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV127955.1 | - | 214 | Chr. - - [-] |