Gene Transcripts

Gene Symbol
Dmrtc1b
Entrez Gene ID
632708
Gene Name
DMRT-like family C1b
Gene Aliases
Dmrt8.2, Gm714
Chromosome Location
Chr. X: 102707881 - 102715354 on Build GRCm38
UniGene ID
Mm.297715
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039116.2 NP_001034205.2 2 153
GenBank mRNA BC150829.1 - 2 153
DQ316775.1 - 1 130
DQ316776.1 - 1 290
DQ316777.1 - 2 208

View Full Product Details