Gene Transcripts

Gene Symbol
Skint10
Entrez Gene ID
230613
Gene Name
selection and upkeep of intraepithelial T cells 10
Gene Aliases
A030001H23Rik, Gm570
Chromosome Location
Chr. 4: 112710830 - 112774861 on Build GRCm38
UniGene ID
Mm.122553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177668.2 NP_808336.1 3 358
GenBank mRNA AK037152.1 - 3 323
EF494905.1 - 3 321
EF494906.1 - 3 321

View Full Product Details