Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164156.1 | NP_001157628.1 | 12 | 1253 | |
NM_016766.3 | NP_058046.2 | 13 | 1373 | ||
GenBank mRNA | AF015309.1 | - | 13 | 1323 | |
AK088116.1 | - | 13 | 1338 | ||
AK158347.1 | - | 13 | 1353 | ||
AK158611.1 | - | 9 | 980 | ||
AK167242.1 | - | 12 | 1213 | ||
AK187767.1 | - | 1 | 16 | ||
BC003746.1 | - | 13 | 1357 | 3486681 | |
BC085099.1 | - | 13 | 1341 | 6391692 | |
BC108340.1 | - | 13 | 1348 | 4504579 |