Gene Transcripts
- Gene Symbol
- Cdc26
- Entrez Gene ID
- 66440
- Gene Name
- cell division cycle 26
- Gene Aliases
- 2010012C09Rik
- Chromosome Location
- Chr. 4: 62393812 - 62408640 on Build GRCm38
- UniGene ID
- Mm.109930
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008199.1 |
- |
1 |
907 |
|
AK047055.1 |
- |
|
1444 |
|
View Full Product Details