Gene Transcripts

Gene Symbol
Mrgpra9
Entrez Gene ID
668725
Gene Name
MAS-related GPR, member A9
Gene Aliases
EG668725, MrgA9
Chromosome Location
Chr. 7: 47234919 - 47252848 on Build GRCm38
UniGene ID
Mm.436563
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288801.1 NP_001275730.1 2 379

View Full Product Details