Gene Transcripts

Gene Symbol
Mrgpra2a
Entrez Gene ID
668727
Gene Name
MAS-related GPR, member A2A
Gene Aliases
Chromosome Location
Chr. 7: 47426325 - 47452167 on Build GRCm38
UniGene ID
Mm.431311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172588.1 NP_001166059.1 411
GenBank mRNA BC064040.1 - 411

View Full Product Details