Gene Transcripts

Gene Symbol
CXCL12
Entrez Gene ID
6387
Gene Name
C-X-C motif chemokine ligand 12
Gene Aliases
IRH, PBSF, SCYB12, SDF1, TLSF, TPAR1
Chromosome Location
Chr. 10: 44292088 - 44385097 on Build GRCh38
UniGene ID
Hs.522891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000609.6 NP_000600.1 2 191
NM_001033886.2 NP_001029058.1 2 191
NM_001178134.1 NP_001171605.1 2 191
NM_001277990.1 NP_001264919.1 2 191
NM_199168.3 NP_954637.1 2 191
GenBank mRNA AK292628.1 - 2 194
AK311814.1 - 2 189
AU120056.1 - 2 186
AY644456.1 - 2 148
AY874118.1 - 2 178
BC039893.1 - 2 169 5729604
CD172464.1 - 2 189
CR450283.1 - 2 99
DA266752.1 - 2 192
DQ345517.1 - 2 143
DQ345518.1 - 2 138
DQ345519.1 - 2 143
DQ345520.1 - 2 143
L36033.1 - 2 178
L36034.1 - 2 178
U16752.1 - 2 179

View Full Product Details