Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136026.2 | NP_001129498.1 | 7 | 850 | |
NM_001136232.2 | NP_001129704.1 | 6 | 704 | ||
NM_030673.3 | NP_109598.2 | 7 | 706 | ||
NM_183352.2 | NP_899195.1 | 6 | 672 | ||
XM_005265379.2 | XP_005265436.1 | 7 | 723 | ||
XM_017007019.1 | XP_016862508.1 | 7 | 760 | ||
XM_017007020.1 | XP_016862509.1 | 6 | 567 | ||
XM_017007021.1 | XP_016862510.1 | 6 | 577 | ||
GenBank mRNA | AB209554.1 | - | 5 | 519 | |
AF052155.1 | - | 6 | 704 | ||
AK095629.1 | - | 4 | 2531 | ||
AK223019.1 | - | 6 | 557 | ||
AK301999.1 | - | 7 | 774 | ||
BC002634.2 | - | 7 | 587 | 3605545 | |
BC006167.1 | - | 3 | 364 | ||
BC091506.1 | - | 6 | 582 | 30408210 | |
CN344033.1 | - | 6 | 684 | ||
L09260.1 | - | 6 | 549 |