Gene Transcripts

Gene Symbol
ALOX5
Entrez Gene ID
240
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
5-LO, 5-LOX, 5LPG, LOG5
Chromosome Location
Chr. 10: 45374166 - 45446121 on Build GRCh38
UniGene ID
Hs.89499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000698.4 NP_000689.1 3 509
NM_001256153.2 NP_001243082.1 3 509
NM_001256154.2 NP_001243083.1 3 509
NM_001320861.1 NP_001307790.1 3 509
NM_001320862.1 NP_001307791.1 3 547
GenBank mRNA AK225603.1 - 3 462
AK315223.1 - 3 459
BC034464.1 - 3 448
BC040450.1 - 3 438
BC130332.1 - 3 462 40146363
BC132677.1 - 3 462 40146699
BC143985.1 - 3 462 9052503
HM592258.1 - 3 395
HM592259.1 - 3 395
HM592260.1 - 3 395
HM592261.1 - 3 395
J03571.1 - 3 429
J03600.1 - 3 439

View Full Product Details