Gene Transcripts
- Gene Symbol
- SGTA
- Entrez Gene ID
- 6449
- Gene Name
- small glutamine rich tetratricopeptide repeat containing alpha
- Gene Aliases
- SGT, alphaSGT, hSGT
- Chromosome Location
- Chr. 19: 2754714 - 2783356 on Build GRCh38
- UniGene ID
- Hs.203910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003021.3 |
NP_003012.1
|
8, 9 |
855 |
|
XM_011528178.2 |
XP_011526480.1
|
8, 9 |
909 |
|
GenBank mRNA |
AF368279.1 |
- |
7, 8 |
729 |
|
AF408399.1 |
- |
7, 8 |
708 |
|
AJ133129.1 |
- |
8, 9 |
750 |
|
AJ223828.1 |
- |
7, 8 |
749 |
|
AK293535.1 |
- |
8, 9 |
826 |
|
AL050156.1 |
- |
8, 9 |
817 |
|
BC000390.2 |
- |
8, 9 |
758 |
2821268 |
BC002989.1 |
- |
8, 9 |
758 |
2821268 |
BC005165.2 |
- |
8, 9 |
752 |
3532317 |
BC008885.2 |
- |
8, 9 |
752 |
4302652 |
CR533517.1 |
- |
7, 8 |
708 |
|
CR542282.1 |
- |
7, 8 |
708 |
|
View Full Product Details