Gene Transcripts
- Gene Symbol
- SKP1
- Entrez Gene ID
- 6500
- Gene Name
- S-phase kinase-associated protein 1
- Gene Aliases
- EMC19, OCP-II, OCP2, SKP1A, TCEB1L, p19A
- Chromosome Location
- Chr. 5: 134156391 - 134177033 on Build GRCh38
- UniGene ID
- Hs.171626
- Species
- Homo sapiens
- Gene Symbol
- SKP1P2
- Entrez Gene ID
- 728622
- Gene Name
- S-phase kinase-associated protein 1 pseudogene 2
- Gene Aliases
- SKP1B, p19B
- Chromosome Location
- Chr. 12: 16988747 - 16990628 on Build GRCh38
- UniGene ID
- Hs.741380
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036619.1 |
|
1 |
679 |
|
GenBank mRNA |
AK310444.1 |
- |
1 |
679 |
|
View Full Product Details