Gene Transcripts
- Gene Symbol
- Cask
- Entrez Gene ID
- 29647
- Gene Name
- calcium/calmodulin-dependent serine protein kinase (MAGUK family)
- Gene Aliases
- Chromosome Location
- Chr. X: 9815652 - 10156155 on Build Rnor_6.0
- UniGene ID
- Rn.72627
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022184.1 |
NP_071520.1
|
9 |
1202 |
|
GenBank mRNA |
U47110.1 |
- |
9 |
1202 |
|
View Full Product Details