Gene Transcripts

Gene Symbol
SNAPC1
Entrez Gene ID
6617
Gene Name
small nuclear RNA activating complex polypeptide 1
Gene Aliases
PTFgamma, SNAP43
Chromosome Location
Chr. 14: 61762357 - 61796428 on Build GRCh38
UniGene ID
Hs.179312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003082.3 NP_003073.1 5 729
GenBank mRNA AK314933.1 - 5 729
BC014984.1 - 5 637 4872279
BC019038.2 - 5 633 4590014
U44754.1 - 5 637
Z47542.1 - 5 649

View Full Product Details