Gene Transcripts

Gene Symbol
Ifit1
Entrez Gene ID
56824
Gene Name
interferon-induced protein with tetratricopeptide repeats 1
Gene Aliases
Garg16
Chromosome Location
Chr. 1: 252944105 - 252946170 on Build Rnor_6.0
UniGene ID
Rn.48677
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020096.1 NP_064481.1 1 927
GenBank mRNA AJ276893.1 - 1 927

View Full Product Details