Gene Transcripts

Gene Symbol
TROVE2
Entrez Gene ID
6738
Gene Name
TROVE domain family member 2
Gene Aliases
RO60, RORNP, SSA2
Chromosome Location
Chr. 1: 193059422 - 193091777 on Build GRCh38
UniGene ID
Hs.288178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042369.2 NP_001035828.1 5 1357
NM_001042370.2 NP_001035829.2 5 1357
NM_001173524.1 NP_001166995.1 5 1363
NM_001173525.1 NP_001166996.1 5 1357
NM_004600.5 NP_004591.2 5 1357
NR_033393.1 4 762
XM_006711495.3 XP_006711558.1 6 1429
XM_006711496.3 XP_006711559.1 7 1605
XM_006711497.3 XP_006711560.1 6 1539
XM_011509922.2 XP_011508224.1 5 938
XM_017002180.1 XP_016857669.1 6 1238
XM_017002181.1 XP_016857670.1 6 1467
XM_017002182.1 XP_016857671.1 5 1172
XM_017002183.1 XP_016857672.1 3 509
GenBank mRNA AK314594.1 - 5 1150
AY205314.1 - 5 1171
AY205315.1 - 5 1171
BC036658.2 - 5 1199 4821776
BX394323.2 - 4 335
CR936683.1 - 4 731
J04137.1 - 4 1026
M25077.1 - 5 1171

View Full Product Details