Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001051.4 | NP_001042.1 | 2 | 1454 | |
NM_001278687.2 | NP_001265616.1 | 2 | 1047 | ||
XM_005261721.4 | XP_005261778.1 | 2 | 947 | ||
XM_006724311.2 | XP_006724374.1 | 3 | 1662 | ||
XM_011530349.2 | XP_011528651.1 | 7 | 1797 | ||
XM_017028923.1 | XP_016884412.1 | 4 | 1526 | ||
XM_017028924.1 | XP_016884413.1 | 4 | 2271 | ||
GenBank mRNA | AK291165.1 | - | 2 | 1450 | |
BC042068.1 | - | 2 | 1391 | ||
BC096829.1 | - | 1 | 875 | 6971896 | |
CR456585.1 | - | 2 | 928 |