Gene Transcripts

Gene Symbol
STIM1
Entrez Gene ID
6786
Gene Name
stromal interaction molecule 1
Gene Aliases
D11S4896E, GOK, IMD10, STRMK, TAM, TAM1
Chromosome Location
Chr. 11: 3855703 - 4093210 on Build GRCh38
UniGene ID
Hs.501735
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277962.1 NP_001264891.1 12, 13 2155
NM_003156.3 NP_003147.2 11, 12 2118
GenBank mRNA AK310510.1 - 8, 9 1244
AK314928.1 - 12, 13 1676
BC021300.2 - 11, 12 2110 4899542
JX014264.1 - 11, 12 1550
U52426.1 - 11, 12 2115

View Full Product Details