Gene Transcripts

Gene Symbol
ANK2
Entrez Gene ID
287
Gene Name
ankyrin 2, neuronal
Gene Aliases
ANK-2, LQT4, brank-2
Chromosome Location
Chr. 4: 112706083 - 113383740 on Build GRCh38
UniGene ID
Hs.620557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127493.1 NP_001120965.1 32 3599
NM_001148.4 NP_001139.3 30 3505
NM_020977.3 NP_066187.2 30 3505
XM_005262945.2 XP_005263002.1 29 3504
XM_017008067.1 XP_016863556.1 32 4462
XM_017008068.1 XP_016863557.1 33 4703
XM_017008069.1 XP_016863558.1 31 4437
XM_017008070.1 XP_016863559.1 32 4648
XM_017008071.1 XP_016863560.1 33 3631
XM_017008072.1 XP_016863561.1 32 3607
XM_017008073.1 XP_016863562.1 30 3603
XM_017008074.1 XP_016863563.1 32 3577
XM_017008075.1 XP_016863564.1 31 3541
XM_017008076.1 XP_016863565.1 30 3603
XM_017008077.1 XP_016863566.1 29 3418
XM_017008078.1 XP_016863567.1 30 3603
XM_017008079.1 XP_016863568.1 28 3333
XM_017008080.1 XP_016863569.1 30 3454
XM_017008081.1 XP_016863570.1 29 3504
XM_017008082.1 XP_016863571.1 31 3681
XM_017008083.1 XP_016863572.1 31 3744
XM_017008084.1 XP_016863573.1 31 3639
XM_017008085.1 XP_016863574.1 31 3600
XM_017008086.1 XP_016863575.1 30 3501
XM_017008087.1 XP_016863576.1 32 3780
XM_017008088.1 XP_016863577.1 33 3574
XM_017008089.1 XP_016863578.1 30 3747
XM_017008090.1 XP_016863579.1 31 3744
XM_017008091.1 XP_016863580.1 29 3441
XM_017008092.1 XP_016863581.1 31 3541
XM_017008093.1 XP_016863582.1 32 3538
XM_017008094.1 XP_016863583.1 31 3600
XM_017008095.1 XP_016863584.1 29 3418
XM_017008096.1 XP_016863585.1 33 3574
XM_017008097.1 XP_016863586.1 30 3379
XM_017008098.1 XP_016863587.1 31 3541
XM_017008099.1 XP_016863588.1 32 3538
XM_017008100.1 XP_016863589.1 29 3343
XM_017008101.1 XP_016863590.1 29 3504
XM_017008102.1 XP_016863591.1 28 3319
XM_017008103.1 XP_016863592.1 31 3600
XM_017008104.1 XP_016863593.1 30 3442
XM_017008105.1 XP_016863594.1 29 3418
XM_017008106.1 XP_016863595.1 28 3405
XM_017008107.1 XP_016863596.1 30 3379
XM_017008108.1 XP_016863597.1 28 3303
XM_017008109.1 XP_016863598.1 29 3403
XM_017008110.1 XP_016863599.1 30 3442
XM_017008111.1 XP_016863600.1 29 3418
XM_017008112.1 XP_016863601.1 27 3220
GenBank mRNA AK294720.1 - 9 1250
AK299767.1 - 31 3529
AK299893.1 - 3 347
AK307601.1 - 30 3456
BX537758.1 - 32 3573
BX538132.1 - 4 442
CN259048.1 - 3 407
X56958.2 - 30 3420
Z26634.2 - 30 3405

View Full Product Details