Gene Transcripts

Gene Symbol
TAF11
Entrez Gene ID
6882
Gene Name
TATA-box binding protein associated factor 11
Gene Aliases
MGC:15243, PRO2134, TAF2I, TAFII28
Chromosome Location
Chr. 6: 34877778 - 34888071 on Build GRCh38
UniGene ID
Hs.112444
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270488.1 NP_001257417.1 2 305
NM_005643.3 NP_005634.1 2 305
XM_011514827.2 XP_011513129.1 2 695
GenBank mRNA AF118094.1 - 2 45
AK302226.1 - 2 276
AK313475.1 - 2 246
BC021972.2 - 2 261 3935882
BT019404.1 - 2 191
BT019405.1 - 2 191
BX538195.1 - 1 102
BX647568.1 - 1 473
CR542068.1 - 2 191
D63705.1 - 2 274
DA153455.1 - 2 305
X83928.1 - 2 283

View Full Product Details