Gene Transcripts

Gene Symbol
TARBP2
Entrez Gene ID
6895
Gene Name
TARBP2, RISC loading complex RNA binding subunit
Gene Aliases
LOQS, TRBP, TRBP1, TRBP2
Chromosome Location
Chr. 12: 53500921 - 53506431 on Build GRCh38
UniGene ID
Hs.326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004178.4 NP_004169.3 7 735
NM_134323.1 NP_599150.1 7 1200
NM_134324.2 NP_599151.2 7 775
XM_005269114.1 XP_005269171.1 7 770
XM_005269115.2 XP_005269172.1 7 777
XM_005269120.4 XP_005269177.1 7 1183
XM_005269122.3 XP_005269179.1 7 755
XM_006719581.1 XP_006719644.1 6 619
XM_011538712.2 XP_011537014.1 7 824
XM_017019911.1 XP_016875400.1 7 913
GenBank mRNA AK290737.1 - 7 773
BC002419.2 - 7 1183
BC005860.2 - 7 1065 2960701
BT007140.1 - 7 712
M60801.1 - 7 703
U08998.2 - 7 800

View Full Product Details