Gene Transcripts

Gene Symbol
Ppat
Entrez Gene ID
117544
Gene Name
phosphoribosyl pyrophosphate amidotransferase
Gene Aliases
Atase
Chromosome Location
Chr. 14: 33580541 - 33614919 on Build Rnor_6.0
UniGene ID
Rn.18690
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057198.2 NP_476546.1 2, 3 765
GenBank mRNA AY724485.1 - 2, 3 659
BC086999.1 - 2, 3 765 7189259
D10853.1 - 2, 3 273

View Full Product Details