Gene Transcripts

Gene Symbol
Bod1l1
Entrez Gene ID
207118
Gene Name
biorientation of chromosomes in cell division 1-like 1
Gene Aliases
Abp-10, Abp10, Bod1l
Chromosome Location
Chr. 14: 73738137 - 73792935 on Build Rnor_6.0
UniGene ID
Rn.202713
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770238.1 XP_008768460.1 10 3679
GenBank mRNA D64062.1 - 1 1450

View Full Product Details