Gene Transcripts

Gene Symbol
TMSB4X
Entrez Gene ID
7114
Gene Name
thymosin beta 4, X-linked
Gene Aliases
FX, PTMB4, TB4X, TMSB4
Chromosome Location
Chr. X: 12975107 - 12977227 on Build GRCh38
UniGene ID
Hs.437277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021109.3 NP_066932.1 1 431
GenBank mRNA AK055976.1 - 2 1505
AK057827.1 - 3 428
AV715954.1 - 3 431
BC001631.1 - 1 412
BC016732.1 - 1 432
BC022857.1 - 1 432
BC061586.1 - 3 418
BC062436.1 - 1 432
BC064954.1 - 3 412
BC070248.1 - 3 429
BC070249.1 - 1 428
BC083509.1 - 1 428
BC092437.1 - 1 428
BC104464.1 - 1 382 40031330
BC104465.1 - 1 382 40031337
BC104666.1 - 3 432
BC107789.1 - 3 435
BC139925.1 - 1 417 40148985
BC151215.1 - 1 418 40148015
M11948.1 - 1 228
M17733.1 - 3 429
X02493.1 - 2 332

View Full Product Details