Gene Transcripts

Gene Symbol
Tp53i13
Entrez Gene ID
287550
Gene Name
tumor protein p53 inducible protein 13
Gene Aliases
RGD1559818, Trp53i13
Chromosome Location
Chr. 10: 62650641 - 62655327 on Build Rnor_6.0
UniGene ID
Rn.17771
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105818.1 NP_001099288.1 4 343
XM_006246902.2 XP_006246964.1 3 370
GenBank mRNA BC158678.1 - 4 305

View Full Product Details