Gene Transcripts

Gene Symbol
Scarf2
Entrez Gene ID
287949
Gene Name
scavenger receptor class F, member 2
Gene Aliases
Chromosome Location
Chr. 11: 87722350 - 87733741 on Build Rnor_6.0
UniGene ID
Rn.22134
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105864.1 NP_001099334.1 10 1629
XM_006248688.2 XP_006248750.1 10 1632

View Full Product Details