Gene Transcripts

Gene Symbol
Mis18a
Entrez Gene ID
288272
Gene Name
MIS18 kinetochore protein A
Gene Aliases
RGD1310778
Chromosome Location
Chr. 11: 30880231 - 30893262 on Build Rnor_6.0
UniGene ID
Rn.137223
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127523.2 NP_001120995.2 3, 4 520
GenBank mRNA BC167102.1 - 3, 4 520

View Full Product Details