Gene Transcripts

Gene Symbol
Gtf2h3
Entrez Gene ID
288651
Gene Name
general transcription factor IIH, polypeptide 3
Gene Aliases
Chromosome Location
Chr. 12: 37427556 - 37444027 on Build Rnor_6.0
UniGene ID
Rn.203542
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024236.1 NP_001019407.1 6, 7 491
GenBank mRNA BC093380.1 - 6, 7 491

View Full Product Details