Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199427.1 | NP_001186356.1 | 4 | 528 | |
NM_003300.3 | NP_003291.2 | 4 | 528 | ||
NM_145725.2 | NP_663777.1 | 5 | 667 | ||
NM_145726.2 | NP_663778.1 | 5 | 667 | ||
XM_011537116.2 | XP_011535418.1 | 5 | 643 | ||
XM_011537117.2 | XP_011535419.1 | 5 | 643 | ||
XM_011537118.2 | XP_011535420.1 | 5 | 644 | ||
XM_017021617.1 | XP_016877106.1 | 5 | 1087 | ||
XM_017021618.1 | XP_016877107.1 | 5 | 2583 | ||
XM_017021619.1 | XP_016877108.1 | 5 | 2585 | ||
XM_017021620.1 | XP_016877109.1 | 5 | 645 | ||
GenBank mRNA | AK303172.1 | - | 4 | 405 | |
BC075086.2 | - | 3 | 339 | 30915387 | |
BC075087.2 | - | 3 | 339 | 30915599 | |
BX247977.1 | - | 5 | 513 | ||
L38509.1 | - | 4 | 450 | ||
U15637.1 | - | 5 | 524 | ||
U19260.1 | - | 4 | 464 | ||
U21092.1 | - | 4 | 531 |