Gene Transcripts

Gene Symbol
Gtpbp3
Entrez Gene ID
290633
Gene Name
GTP binding protein 3
Gene Aliases
Chromosome Location
Chr. 16: 19897135 - 19902226 on Build Rnor_6.0
UniGene ID
Rn.109111
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011919.1 NP_001011919.1 6 843
GenBank mRNA BC087083.1 - 6 843

View Full Product Details