Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_212510.1 | NP_997675.1 | 3 | 292 | |
GenBank mRNA | BC058498.1 | - | 3 | 292 | |
FQ209874.1 | - | 2 | 133 | ||
FQ209881.1 | - | 3 | 286 | ||
FQ209956.1 | - | 3 | 292 | ||
FQ216877.1 | - | 3 | 285 | ||
FQ221165.1 | - | 3 | 287 | ||
FQ221243.1 | - | 3 | 286 | ||
FQ221413.1 | - | 3 | 286 | ||
FQ221458.1 | - | 3 | 286 | ||
FQ221831.1 | - | 3 | 293 | ||
FQ222162.1 | - | 3 | 286 | ||
FQ222289.1 | - | 3 | 286 | ||
FQ222457.1 | - | 3 | 286 | ||
FQ222474.1 | - | 3 | 290 | ||
FQ222662.1 | - | 3 | 286 | ||
FQ223818.1 | - | 3 | 287 | ||
FQ228440.1 | - | 3 | 286 | ||
X14181.1 | - | 3 | 286 |