Gene Transcripts

Gene Symbol
Prtfdc1
Entrez Gene ID
291355
Gene Name
phosphoribosyl transferase domain containing 1
Gene Aliases
Chromosome Location
Chr. 17: 87926242 - 88037051 on Build Rnor_6.0
UniGene ID
Rn.33792
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106127.1 NP_001099597.1 2 113
XM_008771860.1 XP_008770082.1 2 124
GenBank mRNA BC166887.1 - 2 110
BC166923.1 - 2 110 6919422

View Full Product Details