Gene Transcripts

Gene Symbol
Vom1r53
Entrez Gene ID
292607
Gene Name
vomeronasal 1 receptor 53
Gene Aliases
V1rm3
Chromosome Location
Chr. 1: 66397676 - 66398629 on Build Rnor_6.0
UniGene ID
Rn.129414
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008934.1 NP_001008934.1 1 718
GenBank mRNA AY510369.1 - 1 718

View Full Product Details