Gene Transcripts

Gene Symbol
Kif7
Entrez Gene ID
293047
Gene Name
kinesin family member 7
Gene Aliases
Chromosome Location
Chr. 1: 141434183 - 141452578 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229414.2 XP_006229476.1 5 1122
XM_006229415.2 XP_006229477.1 5 1122
XM_008759585.1 XP_008757807.1 5 1122
XM_218828.8 XP_218828.5 5 1122

View Full Product Details