Gene Transcripts

Gene Symbol
Lipk
Entrez Gene ID
294094
Gene Name
lipase, family member K
Gene Aliases
Lipl2
Chromosome Location
Chr. 1: 252323865 - 252355541 on Build Rnor_6.0
UniGene ID
Rn.218534
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106374.1 NP_001099844.1 3 282
XM_008760325.1 XP_008758547.1 4 290

View Full Product Details