Gene Transcripts
- Gene Symbol
- Taar7b
- Entrez Gene ID
- 294126
- Gene Name
- trace amine-associated receptor 7b
- Gene Aliases
- TA12
- Chromosome Location
- Chr. 1: 22518560 - 22519636 on Build Rnor_6.0
- UniGene ID
- Rn.138141
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175586.2 |
NP_783176.2
|
1 |
1035 |
|
- Gene Symbol
- Taar7e
- Entrez Gene ID
- 294133
- Gene Name
- trace-amine-associated receptor 7e
- Gene Aliases
- TA14
- Chromosome Location
- Chr. 1: 22479446 - 22480522 on Build Rnor_6.0
- UniGene ID
- Rn.138144
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175590.2 |
NP_783180.2
|
1 |
1035 |
|
View Full Product Details